Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms
Catalog No. p-6153225
Change view
Click to view available options
Size:
11 mm
13 mm
20 mm
Type:
Manual Crimper
Manual Decrimper
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Type
03-378-609 11 mm Manual Crimper
03-378-611 13 mm Manual Crimper
03-378-612 13 mm Manual Decrimper
03-378-618 20 mm Manual Decrimper
03-378-610 11 mm Manual Decrimper
03-377-446 20 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-378-609 Supplier Thermo Scientific™ Supplier No. C4012100SS
Only null left
Add to Cart
Add to Cart
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
No. per Pack 1 Ea.
Type Manual Crimper
For Use With (Equipment) Aluminum Seal
Size 11 mm
For Use With (Application) Industrial Chromatography
Product Line National
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.