Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard: Caution Biological Hazard Sign
Shop All AccuformNMC Products
Click to view available options
Material:
Plastic
Vinyl
Description
- Orange sign with black text and graphic
- Available in rigid plastic or pressure-sensitive vinyl
- Size: 17.7 × 43.2cm (7 in. × 17 in.)
Specifications
Specifications
Certifications/Compliance | OSHA |
Material | Plastic |
Quantity | 1 Ea. |
Height (English) | 7 in. |
Width (English) | 17 in. |
Displayed Text | BIOHAZARD CAUTION BIOLOGICAL HAZARD/Biohazard Pictogram |
Product Type | Biohazard Sign |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction