Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard: Infectious Waste Sign
Shop All AccuformNMC Products
Click to view available options
Material:
Plastic
Vinyl
Description
- Orange sign with black text
- Available in rigid plastic or pressure-sensitive vinyl
- Dimensions: 25.4 × 35.6cm (10 × 14 in.)
Specifications
Specifications
| Material | Vinyl |
| Quantity | 1 Ea. |
| Height (English) | 10 in. |
| Width (English) | 14 in. |
| Displayed Text | BIOHAZARD INFECTIOUS WASTE |
| Product Type | Biohazard Sign |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction