Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard Signs
Shop All AccuformNMC Products
Click to view available options
Material:
Plastic
Vinyl
Description
- Orange sign with black text
- Available in rigid plastic or pressure-sensitive vinyl Sized 17.7 × 17.7cm (7 × 7 in.)
Specifications
Specifications
| Material | Plastic |
| Quantity | 1 Ea. |
| Height (English) | 7 in. |
| Width (English) | 7 in. |
| Displayed Text | BIOHAZARD |
| Product Type | Biohazard Sign |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction