Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Chemical Hazard Identification Labels

Catalog No. 17985478 Shop All National Marker Company Products
Change view
Click to view available options
Display Text:
Acetic Acid
Acetone
Chlorine
Diesel Fuel
Ethyl Alcohol
Flammable Liquid
Formaldehyde
Gasoline
Hydroxide Peroxide
Isopropyl Alcohol
Mercury
Methyl Alcohol
Nitric Acid
Phosphoric Acid
Propane
Toluene
Xylene
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
17-985-478 Toluene
17-985-422 Acetic Acid
17-985-423 Acetone
17-985-432 Chlorine
17-985-435 Ethyl Alcohol
17-985-441 Formaldehyde
17-985-439 Flammable Liquid
17-985-451 Hydroxide Peroxide
17-985-446 Gasoline
17-985-452 Isopropyl Alcohol
17-985-456 Mercury
19-035-043 Methyl Alcohol
17-985-464 Nitric Acid
17-985-467 Phosphoric Acid
17-985-472 Propane
19-035-039 Diesel Fuel
19-035-046 Xylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 17-985-478 Supplier National Marker™ Supplier No. HC322P

High visibility red-and-white pressure-sensitive vinyl labels ideal for alerting employees to potential chemical hazards in the workplace

  • Made to withstand most chemicals, solvents and vapors
  • Resist fading and scratching
  • For marking larger containers
  • Measure 6 × 10 in.

Specifications

Color White, Red
Format Label
Material Vinyl
Product Type Container Label
Display Text Toluene
For Use With (Application) For marking larger containers, both indoor and outdoor applications
Quantity 1 Ea.
Width (English) 10 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.