Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Chemical Hazard Identification Labels
Click to view available options
Display Text:
Acetic Acid
Acetone
Chlorine
Diesel Fuel
Ethyl Alcohol
Flammable Liquid
Formaldehyde
Gasoline
Hydroxide Peroxide
Isopropyl Alcohol
Mercury
Methyl Alcohol
Nitric Acid
Phosphoric Acid
Propane
Toluene
Xylene
Description
- Made to withstand most chemicals, solvents and vapors
- Resist fading and scratching
- For marking larger containers
- Measure 6 × 10 in.
Specifications
Specifications
Color | White, Red |
Format | Label |
Material | Vinyl |
Product Type | Container Label |
Display Text | Formaldehyde |
For Use With (Application) | For marking larger containers, both indoor and outdoor applications |
Quantity | 1 Ea. |
Width (English) | 10 in. |
Height (English) | 6 in. |
Height (Metric) | 15.24 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction