Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Danger: Permit Required Confined Space Do Not Enter Signs

Catalog No. p-3659886 Shop All AccuformNMC Products
Click to view available options
Dimensions (L x W):
25.4 x 35.6 cm
7 x 10 in.
Material:
Plastic
Vinyl

Catalog No. 18-999-728

Supplier: National Marker™ D360RB

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Material Plastic
Quantity 1 Ea.
Description Rigid Plastic
For Use With (Application) Perfect way to alert employees and guests when a hazard is present in your facility
Height (English) 10 in.
Max. Temperature (English) 183°F
Max. Temperature (Metric) 84°C
Thickness 0.05 in.
Width (English) 14 in.
Dimensions (L x W) 25.4 x 35.6 cm
Displayed Text DANGER Permit Required Confined Space Do Not Enter
Product Type OSHA Safety Sign
Color Red on White
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.