Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Glo-Brite Exit Signs

Catalog No. 19065995 Shop All National Marker Company Products
Change view
Click to view available options
Product Type:
Double-faced Framed Sign
Single-face Framed Sign
Single-faced Framed Sign w/ Bracket
Wall Mount Bracket
Color:
Black
Green
Green Illumination
Red
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
19-065-995 Double-faced Framed Sign Green Illumination
19-065-992 Wall Mount Bracket Black
19-065-993 Single-face Framed Sign Black
19-065-994 Single-faced Framed Sign w/ Bracket Green Illumination
19-065-996 Wall Mount Bracket Red
19-066-001 Single-face Framed Sign Green Illumination
19-065-998 Single-faced Framed Sign w/ Bracket Green Illumination
19-066-002 Single-faced Framed Sign w/ Bracket Green Illumination
19-065-997 Single-face Framed Sign Green Illumination
19-065-999 Double-faced Framed Sign Green Illumination
19-066-004 Double-faced Framed Sign Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19-065-995 Supplier National Marker™ Supplier No. 7002B

Specifications

Certifications/Compliance UL 924, CUL US
Includes Mounting bracket
Material Steel, Plastic
Quantity 1 Ea.
Mounting Type Wall Mount
For Use With (Application) For both high level and low level applications
Height (English) 8.25 in.
Width (English) 15.25 in.
Dimensions (L x W) 8.25 x 15.25 in.
Displayed Text Exit (Left and Right arrow)
Product Type Double-faced Framed Sign
Color Green Illumination
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.