Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Hazardous Material Pressure Sensitive Numbers for Labeling
Click to view available options
Display Text:
1
2
3
4
Specifications
Specifications
Color | Black, White |
Format | Label |
Material | Vinyl |
Product Type | Label Number |
Display Text | 3 |
For Use With (Application) | Hazardous materials |
Quantity | 5/Pk. |
Thickness (English) | 0.0045 mil |
Width (English) | 5 in. |
Height (English) | 5 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction