Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Hazardous Materials Classification Reference Signs
Click to view available options
Material:
Plastic
Vinyl
Width (English):
2.5 in.
8 in.
10 in.
Description
- Easy-to-understand reference signs
- Graphically explain the NFPA hazard identification coding system.
- Available in 0.03 in. thick rigid plastic with predrilled mounting holes, or in 4mil thick pressure-sensitive vinyl
Specifications
Specifications
Material | Vinyl |
Quantity | 1 Ea. |
For Use With (Application) | Training or quick reference, graphically explain the NFPA hazard identification coding system |
Height (English) | 14 in. |
Thickness | 4 mil |
Width (English) | 10 in. |
Displayed Text | Hazardous Materials Classification Reference |
Product Type | Hazardous Materials Classification Reference Sign |
Color | Yellow on White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction