Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Hazardous Materials Classification Reference Signs

Catalog No. p-167266 Shop All AccuformNMC Products
Click to view available options
Material:
Plastic
Vinyl
Width (English):
8 in.

Catalog No. 17-984-4B

Supplier: National Marker™ HMC8R

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Ideal for training or quick reference

  • Easy-to-understand reference signs
  • Graphically explain the NFPA hazard identification coding system.
  • Available in 0.03 in. thick rigid plastic with predrilled mounting holes, or in 4mil thick pressure-sensitive vinyl

Specifications

Material Plastic
Quantity 1 Ea.
Description Rigid Plastic
For Use With (Application) Training or quick reference, graphically explain the NFPA hazard identification coding system
Height (English) 11 in.
Thickness 0.03 in.
Width (English) 8 in.
Displayed Text Hazardous Materials Classification Reference
Product Type Hazardous Materials Classification Reference Sign
Color Blue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.