Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Hazardous Materials Classification Reference Signs

Catalog No. 1798534A Shop All AccuformNMC Products
Click to view available options
Material:
Plastic
Vinyl
Width (English):
2.5 in.
8 in.
10 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for training or quick reference

  • Easy-to-understand reference signs
  • Graphically explain the NFPA hazard identification coding system.
  • Available in 0.03 in. thick rigid plastic with predrilled mounting holes, or in 4mil thick pressure-sensitive vinyl

Specifications

Material Vinyl
Quantity 1 Ea.
For Use With (Application) Training or quick reference, graphically explain the NFPA hazard identification coding system
Height (English) 11 in.
Thickness 4 mil
Width (English) 8 in.
Displayed Text Hazardous Materials Classification Reference
Product Type Hazardous Materials Classification Reference Sign
Color Red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.