Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Vinyl Write-On HMIG Labels

Catalog No. p-189579 Shop All AccuformNMC Products
Click to view available options
Product Type:
Safety Label With symbol squares
Safety Label Without symbol squares

Catalog No. 17-981-61A

Supplier: National Marker™ HM25

Ideal for indoor use

  • Resist water and humidity
  • Printed with fade-resistant UV ink

Order Info

Vinyl labels, sized 6 x 4 in. (15 x 10cm), available with or without squares for personal protection symbols.

Specifications

Certifications/Compliance 29 CFR 1910.1200
Color Red, Yellow, Blue, White
Format Label
Material Vinyl
Product Type Safety Label Without symbol squares
Display Text Health, Flammability, Reactivity, Personal Protection
For Use With (Application) Hazardous materials
Quantity 10/Pk.
Width (English) 4 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.