Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Scientific™ MicroTube™ Rack System

Catalog No. p-4399283
Click to view available options
Sterility:
Non-sterile
Sterile
Quantity:
10/Pk.
120/Pk.
80/Pk.
960/Pk.
Product Type:
12-strip Microtubes, 1.2mL 12-strips
8-strip Microtubes, 1.2mL 8-strips
Microtubes
Racked MicroTubes, 1.2mL 12-strips
Racked MicroTubes, 1.2mL 8-strips
Racked MicroTubes, 1.2mL Singles

Featuring a rack with an 8 x 12 hole-pattern on traditional 9mm centers, the entire System conforms to the SBS footprint/configuration compatible with robotic workstations, multichannel pipettors and auto-sampling instruments. An alpha-numeric, non-reversing rack and lid ensure positive sample identification. The lid, with individual condensation rings above each tube, prevents cross-contamination. The inert polypropylene tubes, rack and lid can be autoclaved, centrifuged at moderate speeds and stacked for long-term, low temperature storage of biological samples. The System is ideal for:

  • Bacterial and hybridoma cell uptake
  • Cell harvesting
  • CMV, HSV, HTLV-lll testing
  • Receptor binding assays
  • RIA/EIA serial dilution procedures
  • Recombinant DNA work
  • Pharmaceutical and human genome studies
  • Sample storage librariesRacks, with SBS footprint, feature 8 x 12 hole pattern on 9mm center. making compatible with robotic workstations & multichannel pipettors.

Specifications

Product Type Microtubes
Description MicroTube Dimensions: 9mm dia., 46mm height
Holds Tubes (1.2 mL)
Material Polypropylene
Sterility Non-sterile
Quantity 960/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.