Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ National™ Target™ Kim-Snap™ Closures

Catalog No. p-2363687
Click to view available options
Color:
Yellow
Material (Liner):
PTFE/Rubber

Preassembled with septa for use with Target Snap/Crimp LoVial™ Vials

  • Effective evaporation resistance—apply with simple thumb pressure
  • Replace aluminum crimp seals and end tedious crimping/de-crimping

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) PTFE/Rubber
Color Yellow
Material Polypropylene
Closure Size 11 mm
For Use With (Equipment) Target Snap/Crimp LoVial Vials
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.