Learn More
Thermo Scientific™ National Unassembled Standard Opening Vial, Cap and Septum Convenience Kits
Catalog No. p-4526358
Click to view available options
Marking Spot:
No
Yes
Septum:
PTFE/Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Color:
Amber
Clear
Color (Seal):
Red
Red/Clear
Red/White
Red/White/Red
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
  • Two-compartment, see-through benchtop storage tray
  • Caps with septa are not attached to vials (packaged in plastic bags)
Includes: 
100 vials and 100 caps with pre-assembled septa
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Color (Seal) Red/White/Red
Description Vials: Clear Glass; Closure: Black; Septa: Red PTFE/White Silicone/Red PTFE
Material (Liner) PTFE/Silicone
Color Clear
Material Glass
Septum PTFE/Silicone/PTFE
Marking Spot No
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.