Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NativePac OBE-1 SEC Column for Online Sample Preparation

Catalog No. 01258266
Change view
Click to view available options
Length (Metric):
50 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
01-258-266 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-258-266 Supplier Thermo Scientific™ Supplier No. 43803052130
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Perform Native-MS analysis of biomacromolecules with the NativePac OBE-1 SEC Column designed for rapid exchange of non-volatile buffer components with a volatile buffer.

The Thermo Scientific™ NativePac OBE-1 Column is a Size Exclusion Chromatography (SEC) column designed for rapid exchange of non-volatile buffer components with a volatile buffer for Native-MS analysis of biomacromolecules. The small pores of the SEC stationary phase maximize separation of the non-volatile components from the sample of interest while the short column length minimizes analysis times to less than 2 minutes. A narrow column ID enables the use of low flow rates for high sensitivity detection and maximizes up time by minimizing buffer consumption.

The NativePac OBE-1 SEC column for online sample preparation offers the following features:

  • OBE column for protein molecular weight and structure screening
  • Rapid exchange of non-volatile buffer components with a volatile buffer for Native-MS analysis
  • Short column length minimizes analysis times and buffer consumption
  • Narrow column ID enables low flow rates for high sensitivity
  • Small SEC stationary phase pore size maximizes separation of non-volatile components from samples of interest
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Max. Pressure 1450 psi (100 bar)
Particle Shape Spherical
Particle Size 3 μm
pH 2 to 8
Pore Size 80 Å
Stationary Phase Proprietary Diol
Packing Material Spherical, Fully Porous, Ultrapure Silica
Column Type Online Buffer Exchange
Phase Size Exclusion
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.