Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Natriuretic Peptide Receptor C Antibody (1060830), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB102331K
Description
Natriuretic Peptide Receptor C Monoclonal antibody specifically detects Natriuretic Peptide Receptor C in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
Natriuretic Peptide Receptor C | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4883 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry | |
1060830 | |
Western Blot, Flow Cytometry | |
ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPRC, NPR-C | |
Human embryonic kidney cell, HEK293-derived human NPRC/NPR3, Met1-Glu481, Accession # P17342 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction