Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Natriuretic Peptide Receptor C Antibody (OTI4C3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272881F
Description
Natriuretic Peptide Receptor C Monoclonal specifically detects Natriuretic Peptide Receptor C in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Natriuretic Peptide Receptor C | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NPR3 | |
Full length recombinant protein of human Natriuretic Peptide Receptor C (NP_000899) produced in HEK293T cells. | |
0.1 mL | |
4883.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4C3 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPRC, NPR-C | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction