Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Natriuretic Peptide Receptor C Antibody (SR2154), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32190750UL
Description
Natriuretic Peptide Receptor C Monoclonal antibody specifically detects Natriuretic Peptide Receptor C in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
Natriuretic Peptide Receptor C | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
4883 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Flow Cytometry | |
SR2154 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100 | |
ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPRC, NPR-C | |
A synthesized peptide derived from human Natriuretic Peptide Receptor C (Uniprot #: P17342) | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction