Learn More
ZeptoMetrix NATtrol™ CT/NG External Run Controls

Shop All ZeptoMetrix Corporation Products


Description
- External run controls formulated with purified, intact and chemically modified organisms
- Non infectious and refrigerator stable
- Designed as full process extraction and amplification controls to validate molecular testing
- Supplied in purified protein matrix that mimics composition of true clinical specimen
- Evaluates performance of nucleic acid test
- 6 x 1ml vials (each control)
For verification of clinical assays, development of diagnostic tests and training of laboratory personnel
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Certifications/Compliance | ISO, 13485 |
Content And Storage | 2°C to 8°C |
Format | Neisseria gonorrhoea detected |
Media Type | Vial |
Product Type | NATtrol External Run Control |
Formulation | Formulated with purified, intact bacterial cells (Neisseria gonorrhoea) |
For Use With (Application) | Quality control of clinical assays and training of laboratory personnel |
Quantity | 6 x 1 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.