Learn More
Zeptometrix NATtrol™ CT/NG/TV Controls
Designed to evaluate the performance of nucleic acid tests for determination of the presence of C. trachomatis, N. gonorrhoeae, and T. vaginalis DNA.
$112.00 - $420.00
Specifications
For Use With (Application) | Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance |
---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
22-158-434
![]() |
Zeptometrix
NATCTNGTVNEG |
Negative Controls |
Each for $112.00
|
|
|||||
22-158-433
![]() |
Zeptometrix
NATCTNGTVPOS |
Positive Controls |
Each for $420.00
|
|
|||||
Description
NATtrol™ CT/NG/TV Positive Control is formulated with purified, intact microorganisms. The microorganisms have been chemically modified to render them non-infectious and refrigerator stable. NATtrol™ CT/NG/TV Negative Control is formulated in the absence of any microorganisms. Each control pack contains 6 x 1.0 mL of NATtrol™ CT/NG/TV Positive Control or NATtrol™ CT/NG/TV Negative Control. These controls are provided in a proprietary matrix.
INTENDED USE:
- NATtrol™ CT/NG/TV Positive Control and NATtrol™ CT/NG/TV Negative Control are designed to evaluate the performance of nucleic acid tests for determination of the presence of C. trachomatis, N. gonorrhoeae, and T. vaginalis DNA.
- NATtrol™ CT/NG/TV Positive Control and NATtrol™ CT/NG/TV Negative Control enable laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
Specifications
Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance |
RUO only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.