Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ Cytomegalovirus AD-169
Shop All ZeptoMetrix Corporation Products

Click to view available options
Concentration:
10000 copies/mL
100000 copies/mL
400 copies/mL
5000 copies/mL
50000 copies/mL
Description
- Each vial contains 1.0 mL of CMV NATtrol™ at target concentrations of 400 to 100,000 copies/mL
- Supplied in purified protein matrix identical to the composition of true clinical specimen
- Used to evaluate the performance of nucleic acid tests for determination of the presence of CMV DNA
Contain intact organisms and should be run in a manner identical to that used for clinical specimens.
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 400 copies/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | NATtrol Virus Strain Control |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of CMV DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction