Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Flu Verification Panel
SDP

Catalog No. 22156739 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
7 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-739 7 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-739 Supplier ZeptoMetrix Supplier No. NATFVPNNS
Add to Cart
Add to Cart

Designed to evaluate the performance of nucleic acid tests for determination of the presence of viral nucleic acids

  • formulated with purified, intact virus particles that have been chemically modified to render them noninfectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Contains 7 x 1mL vials

  • Panel members:
  • Influenza A H1, strain A/New Caledonia/20/99
  • Influenza A H3, strain A/Brisbane/10/07
  • Influenza A 2009 H1N1, strain NY/02/09
  • Influenza B, strain B/Florida/02/06
  • Respiratory Syncytial Virus A
  • Respiratory Syncytial Virus B, strain CH93(18)-18
  • Negative

NATFVP-NNS contains intact organisms and should be run in a manner identical to that used for clinical specimens.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Verification Panel
For Use With (Application) Determination of presence of viral Nucleic Acids
Quantity 7 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.