Learn More
ZeptoMetrix NATtrol Hepatitis C Virus Positive Control
SDP
Catalog No. 21124406 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.25 mL
Catalog No. Quantity
21-124-406 6 x 0.25 mL
1 options
This item is not returnable. View return policy
Catalog No. 21-124-406 Supplier ZeptoMetrix Supplier No. NATHCV6CIVD
Add to Cart
Add to Cart
This item is not returnable. View return policy
A qualitative in-vitro diagnostic external control intended to evaluate test performance of molecular assays which detect Hepatitis C virus nucleic acids.
  • Contains purified, intact hepatitis C viral particles that have been chemically modified to render them non-infectious and refrigerator stable and human A-549 cells
  • Formulated in a proprietary matrix
  • Full process control designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures
Specifications
Product Type Positive Control
Sterility Non-sterile
Quantity 6 x 0.25 mL