Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ HIV-1 External Run Controls, IIIB Strain
Shop All ZeptoMetrix Corporation Products

Click to view available options
Concentration:
1000IU/mL
50000IU/mL
Description
- Formulated with purified, intact virus particles that are chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
- Contains 6 x 1mL vials
- Available concentrations: 1000 or 50,000 IU/mL
NATHIV1-ERCL and NATHIV1-ERCM contain intact virus and should be run in a manner identical to that used for clinical specimens
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 50000IU/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | NATtrol External Run Control |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of HIV-1 RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 6 x 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction