Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ Human Hepatitis C Virus
Shop All ZeptoMetrix Corporation Products

Click to view available options
Concentration:
100000IU/mL
10000IU/mL
1000IU/mL
200IU/mL
50000IU/mL
Description
- Source material is human
- Prepared from highly purified viruses
- Viral particles are treated chemically and enzymatically treated to alter its surface proteins and unable to bind, penetrate or infect host cell
- Retains complete viral genome in its core
Used as controls and calibrators for PCR, TMA, Branched Chain DNA and other NAT assays
Specifications
Specifications
Certifications/Compliance | ISO, 13485 |
Concentration | 100000IU/mL |
Content And Storage | 2°C to 8°C |
Form | Solution |
Media Type | Vial |
Product Type | NATtrol Virus Stock |
Formulation | Formulated with purified, intact virus particles |
For Use With (Application) | Determination of presence of HCV RNA, for use as control and calibrators for PCR, TMA, Branched Chain DNA and other NAT assays |
Quantity | 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction