Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Varicella-Zoster Virus, Strain: Ellen
SDP

Catalog No. 22156762 Shop All ZeptoMetrix Corporation Products
Click to view available options
Concentration:
100 copies/mL
1000 copies/mL
10000 copies/mL
500 copies/mL
50000 copies/mL

Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable

  • Each vial contains 1.0mL of VZV NATtrol at various concentrations
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Can be used to evaluate the performance of nucleic acid tests for determination of the presence of VZV DNA

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Concentration 500 copies/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Virus Strain Control
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of VZV RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.