Learn More
ZeptoMetrix NATtrol™ Zika Virus, External Run Control, Low
Shop All ZeptoMetrix Corporation Products
Click to view available options
Quantity:
6 x 1 mL
Description
- Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
- Can be used for verification of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Quantity | 6 x 1 mL |
Do not use in humans. For research use only. Not for use in diagnostic procedures.