Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Natural Killer Cell Antibody (H25), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250526AF532
Description
Natural Killer Cell Monoclonal specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry.Specifications
Natural Killer Cell | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A purified | |
Immunology | |
Human | |
IgG1 |
Immunohistochemistry | |
H25 | |
Immunohistochemistry | |
NK Cell | |
HSE-2 cell line | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction