Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Natural Killer Cell Antibody (H25), mFluor Violet 500 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250526MFV500
Description
Natural Killer Cell Monoclonal antibody specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Radioimmune AssaysSpecifications
| Natural Killer Cell | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified | 
| Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA) | |
| H25 | |
| Immunohistochemistry, Immunoprecipitation, Radioimmunoassay | |
| NK Cell | |
| HSE-2 cell line | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            