Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Natural Killer Cell Antibody (H25), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250526AF750
Description
Natural Killer Cell Monoclonal specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry.Specifications
| Natural Killer Cell | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Immunology | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| H25 | |
| Immunohistochemistry | |
| NK Cell | |
| HSE-2 cell line | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction