Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nav1.4 Na+ channel Mouse anti-Rat, Unconjugated, Clone: N255/38 , MilliporeSigma™

Catalog No. MABN18511MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN18511MI 100 μg
MABN18512MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN18511MI Supplier MilliporeSigma Supplier No. MABN1851100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nav1.4 Na+ channel Monoclonal antibody specifically detects Nav1.4 Na+ channel in Rat samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nav1.4 Na+ channel
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone N255/38
Conjugate Unconjugated
Gene Accession No. NP_037310
Gene Alias Sodium channel protein type 4 subunit alpha;Mu-1;SkM1;Sodium channel protein skeletal muscle subunit alpha;Sodium channel protein type IV subunit alpha;Voltage-gated sodium channel subunit alpha Nav1.4
Gene Symbols Scn4a
Host Species Mouse
Immunogen Recombinant fragment corresponding to 118 amino acids from the N-terminal half of Rat Voltage-gated sodium channel subunit alpha (Nav1.4).
Molecular Weight of Antigen 208.8 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone N255/38 is a mouse monoclonal antibody that targets Sodium channel protein type 4 subunit alpha (Nav1.4). It targets an epitope within the N-terminal, extracellular domain.
Target Species Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.