Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nav1.7 Mouse anti-Human, Clone: 1D6, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275583
Description
Nav1.7 Monoclonal specifically detects Nav1.7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Nav1.7 | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
ETHA, FEB3B, NE-NA, NENAVoltage-gated sodium channel subunit alpha Nav1.7, Neuroendocrine sodium channel, Peripheral sodium channel 1, PN1hNE-Na, sodium channel protein type 9 subunit alpha, Sodium channel protein type IX subunit alpha, sodium channel, voltage-gated, type IX, alpha subunit, voltage-gated sodium channel alpha subunit Nav1.7, voltage-gated, type IX, alpha polypeptide | |
Mouse | |
226 kDa | |
100 μL | |
6335.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1D6 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide | |
SCN9A | |
Synthetic peptide within Human Nav1.7 aa 1,577-1,626/ 1,988. (SwissProt: Q15858 Human) | |
Protein A purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction