Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nav1.8 Antibody (S134), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147615IR
Description
Nav1.8 Monoclonal specifically detects Nav1.8 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Nav1.8 | |
Monoclonal | |
DyLight 755 | |
hPN3, Nav1.8, PN3, sodium channel protein type 10 subunit alpha, sodium channel protein type X subunit alpha, sodium channel, voltage-gated, type X, alpha subunit, voltage-gated, type X, alpha polypeptide | |
Mouse | |
Protein G purified | |
RUO | |
6336 | |
Human, Mouse, Rat, Monkey | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
S134-12 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
SCN10A | |
Fusion protein amino acids 1724-1956 of rat Nav1.8 | |
0.1 mL | |
Primary | |
Detects approx 220kDa. No cross reactivity against other Nav channels. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction