Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nav1.8 Na+ Channel Monoclonal Antibody (S134-12), QED Bioscience™
SDP

Catalog No. 502277436
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7436 Supplier Leinco Technologies Inc Supplier No. 11523100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nav1.8 Na+ Channel Monoclonal antibody specifically detects Nav1.8 Na+ Channel in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Nav1.8 Na+ Channel
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S134-12
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Scn10a
Gene Accession No. Q62968, Q6QIY3, Q9Y5Y9
Gene Alias FEPS2; hPN3; mPN3; Na(V)1.8; Nav1.8; peripheral nerve sodium channel 3; PN3; Scn10a; Sensory neuron sodium channel; SNS; Sodium channel protein type 10 subunit alpha; sodium channel protein type X subunit alpha; sodium channel type X alpha polypeptide; sodium channel voltage-gated type X alpha polypeptide; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage-gated, type 10, alpha polypeptide; sodium channel, voltage-gated, type X, alpha; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; sodium voltage-gated channel alpha subunit 10; voltage gated sodium channel; voltage-gated sodium channel subunit alpha Nav1.8
Host Species Mouse
Immunogen Fusion protein, aa 1724-1956 (cytoplasmic C-terminus) of rat Nav1.8 (accession no. Q62968).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20264, 29571, 6336
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.