Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAV1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568248
Description
Predicted to react with Human and Mouse samples.
NAV1 belongs to the neuron navigator family and is expressed predominantly in the nervous system. The encoded protein contains coiled-coil domains and a conserved AAA domain characteristic for ATPases associated with a variety of cellular activities. This gene is similar to unc-53, a Caenorhabditis elegans gene involved in axon guidance. The exact function of this gene is not known, but it is thought to play a role in in neuronal development and regeneration. Alternative splicing results in multiple transcript variants.
Specifications
NAV1 | |
Polyclonal | |
Unconjugated | |
Nav1 | |
9530089B19; 9930003A20Rik; C230080M11Rik; KIAA1151; KIAA1213; LOC685707; mKIAA1151; Nav1; Neuron navigator 1; neuron navigator-1; POMFIL3; pore membrane and/or filament interacting like protein 3; pore membrane and/or filament-interacting-like protein 3; similar to neuron navigator 1; STEERIN1; steerin-1; unc-53 homolog 1; Unc53h1 | |
Rabbit | |
affinity chromatography | |
RUO | |
215690, 89796 | |
-20°C | |
Liquid |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.01% sodium azide; pH 7.3 | |
Q8CH77, Q8NEY1 | |
Nav1 | |
Synthesized peptide derived from human NAV1. | |
100 μL | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction