Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAV2 Monoclonal Antibody (4D11)
GREENER_CHOICE

Catalog No. PIMA522827
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522827 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522827 Supplier Invitrogen™ Supplier No. MA522827
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: TGSTPLLRNS HSNSLISECM DSEAETVMQL RNELRDKEMK LTDIRLEALS SAHQLDQLRE AMNRMQSEIE KLKAENDRLK SESQGSGCSR APSQVS.

The vitamin A metabolite, all-trans retinoic acid (atRA), plays an important role in neuronal development, including neurite outgrowth. RAINB1 is an atRA-responsive gene. [supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAV2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene NAV2
Gene Accession No. Q8IVL1
Gene Alias 5330421F07Rik; AU067780; BQ960892; HELAD1; Helicase APC down-regulated 1; helicase, APC down-regulated 1; KIAA1419; mKIAA3015; NAV2; Neuron navigator 2; POMFIL2; Pore membrane and/or filament-interacting-like protein 2; RAINB1; RAINB2; retinoic acid inducible gene in neuroblastoma 1; retinoic acid inducible in neuroblastoma; Retinoic acid inducible in neuroblastoma 1; STEERIN2; Steerin-2; Unc-53 homolog 2; UNC53H2
Gene Symbols NAV2
Host Species Mouse
Immunogen NAV2 (NP_892009.2, 1878 a.a. ~ 1973 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89797
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.