Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAV2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552851 Supplier Invitrogen™ Supplier No. PA552851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTSSSISSYA NTPASSRKNL DVQTDAEKHS QVERNSLWSG DDVKKSDGGS DSGIKMEPGS KWRRNPSDVS DESDKSTSGK KNPVISQTGS WRRGMTAQVG ITMPRTKASA PAGALKTPGT GKTDDAKVSE KG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

The vitamin A metabolite, all-trans retinoic acid (atRA), plays an important role in neuronal development, including neurite outgrowth. RAINB1 is an atRA-responsive gene. [supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAV2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAV2
Gene Accession No. Q8IVL1
Gene Alias 5330421F07Rik; AU067780; BQ960892; HELAD1; Helicase APC down-regulated 1; helicase, APC down-regulated 1; KIAA1419; mKIAA3015; NAV2; Neuron navigator 2; POMFIL2; Pore membrane and/or filament-interacting-like protein 2; RAINB1; RAINB2; retinoic acid inducible gene in neuroblastoma 1; retinoic acid inducible in neuroblastoma; Retinoic acid inducible in neuroblastoma 1; STEERIN2; Steerin-2; Unc-53 homolog 2; UNC53H2
Gene Symbols NAV2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NAV2. Recombinant protein control fragment (Product #RP-89861).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89797
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.