Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAV2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304796100UL
Description
NAV2 Polyclonal specifically detects NAV2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NAV2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
89797 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
EC 3.6.4.12, FLJ10633, FLJ11030, FLJ23707, HELAD1UNC53H2, Helicase APC down-regulated 1, KIAA1419STEERIN2, neuron navigator 2, POMFIL2helicase, APC down-regulated 1, pore membrane and/or filament interacting like protein 2, Pore membrane and/or filament-interacting-like protein 2, RAINB1FLJ77876, retinoic acid inducible gene in neuroblastoma 1, Retinoic acid inducible in neuroblastoma 1, Steerin-2, Unc-53 homolog 2, unc53H2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1800-1980 of human NAV2 (NP_660093.2). | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction