Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NB-2/Cntn5, Rat, Unlabeled, Clone: 1A6,

Catalog No. MABN877MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN877MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN877MI Supplier MilliporeSigma™ Supplier No. MABN877

Rat Monoclonal Antibody

Contactin-5 (UniProt P68500; also known as Neural recognition molecule NB-2) is encoded by the Cntn5 gene (Gene ID 244682) in murine species. Contactin-5 (NB-2) is a glycosylphosphatidylinositol (GPI)-anchored glycoprotein belonging to a subgroup of the immunoglobulin superfamily of proteins known as contactins. Five of the six contactins, Contactin-1, contactin-2/TAG-1, contactin-3/BIG-1, contactin-4/BIG-2, and contactin-5/NB-2 are expressed by DRG neurons. Of these, contactin-1, TAG-1, and BIG-2, are also expressed by motor neurons. NB-2 plays an important role in synapse formation in the developing auditory system in rodents. Expression of NB-2 is seen in the central auditory pathways just before the onset of hearing in rodents and is localized at glutamatergic synapses in the postnatal auditory brainstem. NB-2-knockout mice exhibit abnormal responses to auditory stimuli in both neuronal excitability and behavior, and have delayed auditory brainstem response waves. In addition, NB-2 deficiency in mice causes a significant reduction in the number of synapses and induces apoptosis in a subset of neurons in the auditory brainstem. NB-2 is produced with an N-terminal signal peptide (a.a. 1-23) and a C-terminal propeptide (a.a. 1072-1098) sequence, the removal of which yields the mature GPI-anchored glycoprotein (a.a. 24-1071) with six C2-type Ig-like domains (a.a. 98-659) and four type-III fibronectin domains (a.a. 672-1066).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NB-2/Cntn5
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1A6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Cntn5
Gene Accession No. P68500
Host Species Rat
Immunogen Human IgG Fc-tagged recombinant mouse NB-2/Cntn5.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001164258
Test Specificity Clone 1A6 detected NB-2 in the synaptosomal, but not mitochondrial, fraction from mouse cerebral cortex homogenate (Shimoda, Y., et al. (2012). Neurosci. Lett. 510(2):148-153).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.