Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBAS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103963 Supplier Invitrogen™ Supplier No. PA5103963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NBAS.

Negatively regulates nitric oxide production by inducing NOS1 and NOS3 translocation to actin cytoskeleton and inhibiting their enzymatic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBAS
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NBAS
Gene Accession No. A2RRP1
Gene Alias 4933425L03Rik; ILFS2; NAG; NAG/BC035112 fusion; NAG/FAM49A fusion; NBAS; neuroblastoma amplified sequence; neuroblastoma-amplified gene protein; neuroblastoma-amplified protein; neuroblastoma-amplified protein-like; neuroblastoma-amplified sequence; similar to neuroblastoma-amplified protein; SOPH
Gene Symbols NBAS
Host Species Rabbit
Immunogen A synthesized peptide derived from human NBAS(Accession A2RRP1), corresponding to amino acid residues V1906-N1956.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51594
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.