Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NBAS Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | NBAS |
---|---|
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-0917
![]() |
Bethyl Laboratories, Inc
A305636AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein with two leucine zipper domains, a ribosomal protein S14 signature domain and a Sec39 like domain. The protein is thought to be involved in Golgi-to-ER transport. Mutations in this gene are associated with short stature, optic nerve atrophy, and Pelger-Huet anomaly. [provided by RefSeq, Oct 2012] Involved in Golgi-to-endoplasmic reticulum (ER) retrograde tranport; the function is proposed to depend on its association in the NRZ complex which is believed to play a role in SNARE assembly at the ER (PubMed:19369418).Specifications
NBAS | |
Polyclonal | |
Liquid | |
RUO | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Neuroblastoma-amplified sequence, Neuroblastoma-amplified gene protein, NBAS, NAG | |
NBAS | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
A2RRP1 | |
51594 | |
Between 350 and 400 | |
Primary | |
4° C | |
NBAS |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title