Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBEA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559156 Supplier Invitrogen™ Supplier No. PA559156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKIHTTSDGM SSISERDLAS STKGLEYAEM TATTLETESS SSKIVPNIDA GSIISDTERS DDGKESGKEI RKIQTTTTTQ AVQGRSITQQ DRDLRVDLGF R Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 26%.

This gene encodes a member of a large, diverse group of A-kinase anchor proteins that target the activity of protein kinase A to specific subcellular sites by binding to its type II regulatory subunits. Brain-specific expression and coat protein-like membrane recruitment of a highly similar protein in mouse suggest an involvement in neuronal post-Golgi membrane traffic. Mutations in this gene may be associated with a form of autism. This gene and its expression are frequently disrupted in patients with multiple myeloma. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants may exist, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBEA
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NBEA
Gene Accession No. Q8NFP9
Gene Alias BCL8B; FLJ10197; KIAA1544; Lysosomal-trafficking regulator 2; LYST2; NBEA; Neurobeachin; Protein BCL8B; RP11-270C18.1
Gene Symbols NBEA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NBEA. Recombinant protein control fragment (Product #RP-97999).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.