Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NBEA Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518216
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
This gene encodes a member of a large, diverse group of A-kinase anchor proteins that target the activity of protein kinase A to specific subcellular sites by binding to its type II regulatory subunits. Brain-specific expression and coat protein-like membrane recruitment of a highly similar protein in mouse suggest an involvement in neuronal post-Golgi membrane traffic. Mutations in this gene may be associated with a form of autism. This gene and its expression are frequently disrupted in patients with multiple myeloma. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants may exist, but their full-length nature has not been determined.
Specifications
NBEA | |
Polyclonal | |
Unconjugated | |
NBEA | |
BCL8B; FLJ10197; KIAA1544; Lysosomal-trafficking regulator 2; LYST2; NBEA; Neurobeachin; Protein BCL8B; RP11-270C18.1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
26960 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8NFP9 | |
NBEA | |
Synthetic peptide sequence (DFNRWHYEHQNRY) corresponding to the C-terminus amino acids of NBEA (aa 2934-2946). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction