Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBEAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561974
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561974 Supplier Invitrogen™ Supplier No. PA561974
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TIPNLAISWE GHIVVYSSTE EKTTLKDKNA LHLFSINGKY LGSQILKEQV SDICIIGEHI VTGSIQGFLS IRDLHSLNLS INPLAMRLPI HCVCVTKEYS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

NBEAL1 (Neurobeachin Like 1) is a Protein Coding gene. Diseases associated with NBEAL1 include Ovary Serous Adenocarcinoma. Gene Ontology (GO) annotations related to this gene include binding. An important paralog of this gene is NBEAL2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBEAL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NBEAL1
Gene Accession No. Q6ZS30
Gene Alias A530083I02Rik; ALS2CR16; ALS2CR17; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 16; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 17; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 16 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 17 protein; beach; NBEAL1; neurobeachin like 1; neurobeachin-like 1; neurobeachin-like protein 1
Gene Symbols NBEAL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NBEAL1. Recombinant protein control fragment (Product #RP-99648).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65065
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.