Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110726 Supplier Invitrogen™ Supplier No. PA5110726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPINKLALFP DKSAWCEAKN ITQIVGHSGC EAKSIQNRAC LGQCFSYSVP NTFPQSTESL VHCDSCMPAQ SMWEIVTLEC PGHEEVPRVD KLVEKILHCS CQACGKEPSH EGLSVYVQGE DGPGSQPGTH PHPHPHPHPG GQTPEPE Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.

This gene product is the founding member of the evolutionarily conserved CAN family of proteins, which contain a domain resembling the CTCK motif found in a number of signaling molecules. These proteins are secreted, and act as BMP antagonists by binding to BMPs and preventing them from interacting with their receptors. They may thus play an important role during growth and development. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nbl1
Gene Accession No. P41271
Gene Alias D1S1733E; D4H1S1733E; DAN; DAN domain family member 1; Dana; DAND1; differential screening-selected gene aberrant in neuroblastoma; N03; NB; Nbl1; neuroblastoma 1, DAN family BMP antagonist; neuroblastoma candidate region, suppression of tumorigenicity 1; neuroblastoma suppressor of tumorigenicity 1; neuroblastoma, suppression of tumorigenicity 1; Neuroblastoma, suppression of tumorigenicity 1 (DNA segment human D1S1733E); NO3; Protein N03; Zinc finger protein DAN
Gene Symbols Nbl1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human NBL1. Recombinant protein control fragment (Product #RP-88778).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4681
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.