Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547972
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547972 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547972 Supplier Invitrogen™ Supplier No. PA547972
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) DAN is observed and less than 1% cross-reactivity with recombinant chicken Caronte, rmGremlin, and recombinant human Cerberus 1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

This gene product is the founding member of the evolutionarily conserved CAN family of proteins, which contain a domain resembling the CTCK motif found in a number of signaling molecules. These proteins are secreted, and act as BMP antagonists by binding to BMPs and preventing them from interacting with their receptors. They may thus play an important role during growth and development. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBL1
Applications Immunohistochemistry, Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Nbl1
Gene Accession No. P41271
Gene Alias D1S1733E; D4H1S1733E; DAN; DAN domain family member 1; Dana; DAND1; differential screening-selected gene aberrant in neuroblastoma; N03; NB; Nbl1; neuroblastoma 1, DAN family BMP antagonist; neuroblastoma candidate region, suppression of tumorigenicity 1; neuroblastoma suppressor of tumorigenicity 1; neuroblastoma, suppression of tumorigenicity 1; Neuroblastoma, suppression of tumorigenicity 1 (DNA segment human D1S1733E); NO3; Protein N03; Zinc finger protein DAN
Gene Symbols Nbl1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human DAN Ala17-Asp180 (Ala36Arg).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4681
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.