Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NBN Rabbit anti-Human, Unconjugated, Clone: BGI-14, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20547
Description
NBN Monoclonal antibody specifically detects NBN in Human samples. It is validated for IP, ICC, WB-Ce, IF.Specifications
| NBN | |
| Monoclonal | |
| Unconjugated | |
| NBN | |
| Rabbit | |
| 100 μg | |
| 4683 | |
| Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
| IgG |
| Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
| BGI-14 | |
| nibrin | |
| AT-V1, AT-V2, ATV, FLJ10155, MGC87362, NBS, NBS1, P95 | |
| A synthetic phosphopeptide corresponding to residues surrounding S343 of human NBN | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction