Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NBOH-2C-CN hydrochloride

Catalog No. 517110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-711-0 10 mg
51-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-711-0 Supplier Tocris Bioscience™ Supplier No. 5171/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

High affinity, selective 5-HT2A agonist

High affinity, selective 5-HT2A agonist (Ki = 1.3 nM). Exhibits 100-fold selectivity for 5-HT2A over 5-HT2C. Elicits head twitch behavior and reduces anxiety-related marble-burying activity in mice.

Specifications

Quantity 10 mg
Product Type NBOH-2C-CN hydrochloride
Purity >98%
Molecular Weight (g/mol) 348.82
Inhibitors NBOH-2C-CN hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.